Water/Ice Bath Rack, Electron Microscopy Sciences | Avantor
Water/Ice Bath Rack, Electron Microscopy Sciences
40 Place Rectangular Mailing Rack for 0.5 ml Tubes
Catalog # 100493-768
Fournisseur #:  72380-24
Water/Ice Bath Rack, Electron Microscopy Sciences
Catalog # 100493-768
Supplier:  
Water/Ice Bath Rack, Electron Microscopy Sciences
Water/Ice Bath Rack, Electron Microscopy Sciences
40 Place Rectangular Mailing Rack for 0.5 ml Tubes
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Rust-proof anodized aluminum rack – designed to be used in water/ice baths.
  • Removable cover keeps tubes closed while boiling
  • Handle for easy removal from water baths
  • Will not rust
Spécifications
  • Désignation:
    40 Place Rectangular Mailing Rack for 0.5 ml Tubes