MicroPoint™ Micro Scissors, Electron Microscopy Sciences
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
Featherlite Micro Stork Style Scissors
Catalog # 100492-348
Supplier #:  77902
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
Catalog # 100492-348
Supplier:  
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
MicroPoint™ Micro Scissors, Electron Microscopy Sciences
Featherlite Micro Stork Style Scissors
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Featherlite scissors in different styles.
Style DS-Straight Vannas Scissors, 5 mm and smooth blades. Length 125 mm (5").
Style DS-G – Stork Style, 10 mm long and smooth blades. Length: 125 mm (5").
Style DS-GM – Micro Stork Style, 5 mm long and smooth blades. Length: 125 mm (5").
Specifications
  • Désignation:
    Featherlite Micro Stork Style Scissors