Graduated Pitchers, Electron Microscopy Sciences | Avantor
Graduated Pitchers, Electron Microscopy Sciences
1000 ml
Catalog # 100492-170
Fournisseur #:  74370
Graduated Pitchers, Electron Microscopy Sciences
Catalog # 100492-170
Graduated Pitchers, Electron Microscopy Sciences
Graduated Pitchers, Electron Microscopy Sciences
1000 ml
Price
$44.85
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Gracefully proportioned, molded pitchers with large comfortable handles and well designed pouring spouts.
  • Metric graduations, and crystal clear to the eye (polymethylpentene)
  • Resists heat to 175°C (374°F)
  • Shatter-proof
  • Excellent chemical resistance
Ideal for preparation of photographic chemicals.
Spécifications
  • Capacité:
    1000 ml