Steinheil Magnifier, Electron Microscopy Sciences | Avantor
Steinheil Magnifier, Electron Microscopy Sciences
Steinheil Magnifier, 14×
Catalog # 100491-462
Fournisseur #:  68147
Steinheil Magnifier, Electron Microscopy Sciences
Catalog # 100491-462
Supplier:  
Steinheil Magnifier, Electron Microscopy Sciences
Steinheil Magnifier, Electron Microscopy Sciences
Steinheil Magnifier, 14×
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The magnifier helps the user eliminate bulky and heavier conventional magnifiers.
A biconvex lens. Pocket size. The sliding frame is designed for the storage of the lens after use. It is good for the preliminary inspection of microscopic specimens. It is lightweight and has excellent chromatic correction with a reasonably wide field of view. Size: 39 x 24 x 22 mm. Average weight: 20 g.
Spécifications
  • Grossissement:
    14×
  • Désignation:
    Steinheil Magnifier