Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences | Avantor
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
60 ml (2 oz.)
Catalog # 100490-550
Fournisseur #:  64298-02
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
Catalog # 100490-550
Supplier:  
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
Bellows Dropping Bottles, LDPE, Electron Microscopy Sciences
60 ml (2 oz.)
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This low density polyethylene (LDPE) bottle supplied with a removable bellow-type dropping pipette. The pipettes are graduated for nominal indication of amount of liquid being drawn. Bellows-shaped bulb makes for easy use and provides excellent drop information.
Spécifications
  • Capacité:
    60 ml (2 oz.)