Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences | Avantor
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Med Non-Ferrous, Ti, Cut-Off Wheel
Catalog # 100489-988
Fournisseur #:  50209-25
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Catalog # 100489-988
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Premium Abrasive Cut-off Wheels for Sectioning, Electron Microscopy Sciences
Med Non-Ferrous, Ti, Cut-Off Wheel
Price
$484.51
Log in pour la disponibilité
Each Pack of 5
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Aluminum Oxide and Silicon Carbide Abrasive Cut-Off Wheels are offered for long life, and are affordably priced for various sectioning requirements.
All wheels are 228.6 mm (9") in diameter, 1.6 mm (0.062”) thick, and with 31 mm (1 1/4”) arbor.
Spécifications
  • Désignation:
    Med Non-Ferrous, Ti, Cut-Off Wheel