Frisket Knife, Electron Microscopy Sciences | Avantor
Frisket Knife, Electron Microscopy Sciences
Frisket Knife
Catalog # 100489-648
Fournisseur #:  77618-03
Frisket Knife, Electron Microscopy Sciences
Catalog # 100489-648
Frisket Knife, Electron Microscopy Sciences
Frisket Knife, Electron Microscopy Sciences
Frisket Knife
Price
$30.40
Log in pour la disponibilité
Each Pack of 3
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The frisket Knife is lightweight and disposable. Designed for easy handling.
Comes with a fixed #11 scalpel blade, and a locking safety cap when it is not in use.
Spécifications
  • Désignation:
    Frisket Knife