CD’s Storage Shell and Envelope, Electron Microscopy Sciences
Product Details & Documents
C-Shell™ is designed to provide filling, storage, and protection for compact disks or DVD’s during shipping
Paper Sleeves are economical per sleeve and are far less than plastic jewel case. They are space saving, durable and have clarity.
The C-Shell™ is an ultra slim (only 4.5 mm thick), one-piece polypropylene shell with a living hinge.
Spécifications
- Désignation:Paper Sleeve, CD Storage