Syringes Disposable, Electron Microscopy Sciences
Product Details & Documents
These high quality disposable rubber piston syringes offer substantial savings.
- Lubricated synthetic rubber piston allows for a smooth draw while a positive stop prevents accidental pull out
- Syringes are ETO gas sterilized, latex and pyrogen free
- Bold graduations are easily read and resist wipe-off
Packaging: All syringes are packaged in individual sterile strip packs.
Spécifications
- Capacité:30 ml
- Tip Style:Luerslip Centric