Large Glass Slide Storage Box, Electron Microscopy Sciences
Product Details & Documents
100-Slide Capacity:
These storage boxes are available in two sizes.
100-Slide Capacity:
Durable, wood frame, covered with heavy black embossed paper. The hinges and latches are Nickel plated. A numbered plastic insert provides spacing for the handling of the slides. The box accommodates 100 of 3x2" slides. The box measures 24.7 x 19.7 x 5.7 cm (9 3/4 x 7 4/5 x 2 1/4").
25 Slide Capacity:
Made from wood with numbered partitions which correspond to the index card. Very durable and has a detachable lid. The box accommodates 25 of 3x2" slides.
25 Slide Capacity:
Made from wood with numbered partitions which correspond to the index card. Very durable and has a detachable lid. The box accommodates 25 of 3x2" slides.
Spécifications
- Désignation:100 Microscope Slide Box, 3x2" Slides