Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
Volumetric, 50 ml
  100489-208
 :  70952-50
Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
  100489-208
Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
Graduated Pipettes, Polypropylene, Electron Microscopy Sciences
Volumetric, 50 ml
$596.14
Each Pack of 3
Made from translucent polypropylene.
  • Resistant to breakage
  • Blue graduations, ring mark and inscriptions
Ideal for fluoro and silicate chemistry used. Recommended cleaning with mild alkaline detergent up to 60 °C.
NOTE: Exposure to temperatures above 60 °C may cause volume changes.
  • Capacité:
    50 ml
  • Désignation:
    Volumetric