Zeiss SEM Mounts, Electron Microscopy Sciences
Product Details & Documents
Flat End Pin Surface
- Diameter: 12.7 mm (1/2")
- Slotted Head diameter, 3.1 mm (1/8") Pin
Spécifications
- Désignation:Zeiss Flat End Pin
- Matière:Aluminum