AMRAY 1400 SEM Mounts, Electron Microscopy Sciences
Product Details & Documents
Slotted Head 12.7 mm (1/2") diameter. Pin 3.1 mm (1/8") diameter.
Spécifications
- Désignation:Amray 1400
- Matière:Aluminum