Tungsten Wire Filaments, Electron Microscopy Sciences | Avantor
Tungsten Wire Filaments, Electron Microscopy Sciences
Triple tungsten V-shaped filaments, 5/8" (16 mm) v-opening, 0.5 mm (0.02"), 3"
Catalog # 100488-514
Fournisseur #:  73842
Tungsten Wire Filaments, Electron Microscopy Sciences
Catalog # 100488-514
Tungsten Wire Filaments, Electron Microscopy Sciences
Triple tungsten V-shaped filaments, 5/8" (16 mm) v-opening, 0.5 mm (0.02"), 3"
Price
$97.74
Log in pour la disponibilité
Each Pack of 10
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
V-shaped filaments are high purity, vacuum grade tungsten wire 0.02" (0.50 mm) diameter, V-shaped filaments ½" (12.7mm) deep, with a bottom angle of 45°, either single wire or triple strand.
Coiled filaments are single or 3-strand coiled tungsten filaments, 0.5 mm (0.002") diameter wires.
Spécifications
  • Ø:
    0.5 mm (0.02")
  • Désignation:
    Triple tungsten V-shaped filaments, 5/8" (16 mm) v-opening
  • Longueur:
    3"