Syringes Disposable, Electron Microscopy Sciences | Avantor
Syringes Disposable, Electron Microscopy Sciences
5 ml, Luerslip Centric
Catalog # 100488-480
Fournisseur #:  72509
Syringes Disposable, Electron Microscopy Sciences
Catalog # 100488-480
Syringes Disposable, Electron Microscopy Sciences
Syringes Disposable, Electron Microscopy Sciences
5 ml, Luerslip Centric
Price
$88.59
Log in pour la disponibilité
Each Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These high quality disposable rubber piston syringes offer substantial savings.
  • Lubricated synthetic rubber piston allows for a smooth draw while a positive stop prevents accidental pull out
  • Syringes are ETO gas sterilized, latex and pyrogen free
  • Bold graduations are easily read and resist wipe-off
Packaging: All syringes are packaged in individual sterile strip packs.
Spécifications
  • Capacité:
    5 ml
  • Tip Style:
    Luerslip Centric