High Precision Diamond Scribing Tools, Electron Microscopy Sciences
High Precision Diamond Scribing Tools, Electron Microscopy Sciences
High precision diamond scriber, 0.03" tip, 0.12" tip length
Catalog # 100501-684
Supplier #:  62105-HC
High Precision Diamond Scribing Tools, Electron Microscopy Sciences
Catalog # 100501-684
Supplier:  
High Precision Diamond Scribing Tools, Electron Microscopy Sciences
High Precision Diamond Scribing Tools, Electron Microscopy Sciences
High precision diamond scriber, 0.03" tip, 0.12" tip length
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A newly designed diamond scribing tool set containing eight precision natural diamond scribes in various sizes and configurations. This set is perfect for precision work in microscopy. The set consists of eight diamond tips, 3 mm in diameter and are mounted in interchangeable tool cones and an MT-1 tool handle. The tool and handle are all housed in a hardwood instrument case. Individual scribers can be purchased separately.
Specifications
  • Désignation:
    High precision diamond scriber
  • Longueur:
    0.12" tip length
  • Diameter:
    0.03" tip