Dissecting Dish, Electron Microscopy Sciences | Avantor
Dissecting Dish, Electron Microscopy Sciences
Dissecting Dish with Pyrex Dish
Catalog # 100501-410
Fournisseur #:  70540-50
Dissecting Dish, Electron Microscopy Sciences
Catalog # 100501-410
Dissecting Dish, Electron Microscopy Sciences
Dissecting Dish, Electron Microscopy Sciences
Dissecting Dish with Pyrex Dish
Price
$398.49
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A silicone pad placed inside a 150x20 mm deep plastic petri dish. The mat can be removed, washed, and returned to the dish for reuse. The pins come in 10 different coded colors.
Optional Pyrex Glass Petri dish is available for use when sterilization is needed.
Spécifications
  • Désignation:
    Dissecting Dish with Pyrex Dish