Embedding Rings, Electron Microscopy Sciences | Avantor
Embedding Rings, Electron Microscopy Sciences
Embedding ring, Yellow
Catalog # 100500-980
Fournisseur #:  62350-Y
Embedding Rings, Electron Microscopy Sciences
Catalog # 100500-980
Embedding Rings, Electron Microscopy Sciences
Embedding Rings, Electron Microscopy Sciences
Embedding ring, Yellow
Price
$119.94
Log in pour la disponibilité
Box of 250 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Molded from the highest quality plastic, our Embedding Rings fit most base molds. For added convenience, there are four colors from which to choose Blue, Pink, Green, Yellow. The large writing surface further eases identification and handling. The base measures 1 5/8 x 1 1/8" and the ring measures 25 x 25 mm (1 x 1").
Spécifications
  • Color:
    Yellow
  • Désignation:
    Embedding ring