Transfer Pipette, Polypropylene, Electron Microscopy Sciences | Avantor
Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Transfer Pipettes, Curved, 12 mL
Catalog # 100500-758
Fournisseur #:  70986-20
Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Catalog # 100500-758
Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Transfer Pipette, Polypropylene, Electron Microscopy Sciences
Transfer Pipettes, Curved, 12 mL
Price
$195.51
Log in pour la disponibilité
Box of 50 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Transfer pipette that works as a syringe for liquid transfer. Each pipette has calibration markings from 0 to 12 ml in 0.2 ml increments. Tubing may be attached to the tip to suit your application, and tips may also be cut to facilitate aspiration and delivery of fluids of varying viscosities.
  • Inexpensive and disposable
Spécifications
  • Désignation:
    Transfer Pipettes, Curved, 12 mL