Flat Emery Sticks, Electron Microscopy Sciences
Flat Emery Sticks, Electron Microscopy Sciences
2 grit emery stick
  100499-884
Supplier #:  62119-20
Flat Emery Sticks, Electron Microscopy Sciences
  100499-884
Flat Emery Sticks, Electron Microscopy Sciences
Flat Emery Sticks, Electron Microscopy Sciences
2 grit emery stick
60,12 $
Dozen Each
Emery paper which is glued to a wooden handle. Very useful for filing excess resin of sample block tips. Grit size ranges from number 4/0 (finest) to 3 (coarsest).
Dimensions L x W: 279 x 19.1 mm (11 x 3/4").
  • Désignation:
    2 grit emery stick