Sterilizing Tray Plastic, Electron Microscopy Sciences
Perforated sterilizing tray has a silicone mat which provides flexibility int the layout the instruments for sterilization. A unique "grid" system exists in the base, tray and lid, making it easy to install the mat required, protecting your instruments properly.
- Made of high strength Ultem® Plastic
- Autoclave, Steam, Dry Heat, and Chemical Sterilization are possible
- Has an extended Life Cycle
- Trays are stackable
- Economical and Flexible Design
- Bases have pebble-type surface to keep mats from sticking
The trays are stackable and they have integral feet in base and corner ridges on lids which allow for easier handling in the autoclave and for storage.
- Désignation:Sterilization Tray with Silicone Mat and Lid
- Taille:151 x 64 x 32 mm