Petroff-Hausser Chambers, Electron Microscopy Sciences
Petroff Hausser chamber, 10 µm without grid
Catalog # 100498-486
 :  63512-25
Petroff-Hausser Chambers, Electron Microscopy Sciences
Catalog # 100498-486
Petroff-Hausser Chambers, Electron Microscopy Sciences
Petroff-Hausser Chambers, Electron Microscopy Sciences
Petroff Hausser chamber, 10 µm without grid
Price
$2,944.52
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
New improved Neubauer ruling pattern with 1/400 sq. mm of ruling. Designed for the counting of bacteria, sperm, blood platelets, and the content of vaccines.
Cell is 0.02 mm deep with 1.5 mm thick glass slide allows the use off dark field microscopy.
Spécifications
  • Désignation:
    Petroff Hausser chamber, 10 µm without grid