Swiss Pattern Needle Files, Electron Microscopy Sciences
A 12-piece file set which contains the most popular shapes in a handy plastic pouch.
- Files come with knurled handles
- Excellent quality
- Available in cut #2 only
Blue anodized aluminum handles are well knurled with an excellent grip. The handle features a steel easy-release chuck, which simply presses into the barrel and remains secure.
- Désignation:Small Handle Needle File