PTFE Beakers, Electron Microscopy Sciences
These beakers are statically molded from virgin "PTFE" and machine finished to give a smooth surface for ease of cleaning and to eliminate contamination.
- With pouring spout
- Virtually immune to chemical attack
Working temperature range is from cryogenic to 260°C (500°F). Ideal for use in wafer etching.
- Capacité:1 ml
- Color:White
- Désignation:PTFE Beaker Cover