Brushes, Red Sable, Electron Microscopy Sciences
Product Details & Documents
Extremely soft and pliable brush is perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.
Spécifications
- Bristle Material:Sable hair
- Désignation:Red Sable Brush Set of 3 Sizes (#2/0; #1 and #2)
- Taille:#2/0; #1 and #2