Thin Sample Holder, Electron Microscopy Sciences
Thin Sample Holder, Electron Microscopy Sciences
Thin sample holder, 15 mm pin
Catalog # 100494-358
Supplier #:  75948-15
Thin Sample Holder, Electron Microscopy Sciences
Catalog # 100494-358
Thin Sample Holder, Electron Microscopy Sciences
Thin Sample Holder, Electron Microscopy Sciences
Thin sample holder, 15 mm pin
Price
$146.83
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for examination cross section of thin samples, such as wafers, multi-layer of capacitors, plastics, metals and more.
Specifications
  • Désignation:
    Thin sample holder, 15 mm pin