Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Vannas Scissors Angle Forward, Delicate, 5 mm, Extra fine
Catalog # 100493-892
Supplier #:  72933-03
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Catalog # 100493-892
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Spring Scissors–Micro Scissors, Electron Microscopy Sciences
Vannas Scissors Angle Forward, Delicate, 5 mm, Extra fine
Price
$829.67
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All Vannas micro scissors have extra fine tips and are 3" (75 mm) long. Available with a 3 mm or 5 mm cutting edge.
Specifications
  • Désignation:
    Vannas Scissors Angle Forward, Delicate
  • Blade Length:
    5 mm
  • Blade Type:
    Extra fine