Water/Ice Bath Rack, Electron Microscopy Sciences | Avantor
Water/Ice Bath Rack, Electron Microscopy Sciences
24-place for 1.5/2.0 ml Tubes
Catalog # 100493-766
Fournisseur #:  72380-20
Water/Ice Bath Rack, Electron Microscopy Sciences
Catalog # 100493-766
Water/Ice Bath Rack, Electron Microscopy Sciences
Water/Ice Bath Rack, Electron Microscopy Sciences
24-place for 1.5/2.0 ml Tubes
Price
$403.19
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Rust-proof anodized aluminum rack – designed to be used in water/ice baths.
  • Removable cover keeps tubes closed while boiling
  • Handle for easy removal from water baths
  • Will not rust
Spécifications
  • Désignation:
    24-place for 1.5/2.0 ml Tubes