Safety Bulb, Electron Microscopy Sciences
Safety Bulb
  100491-706
Safety Bulb, Electron Microscopy Sciences
  100491-706
Safety Bulb, Electron Microscopy Sciences
Safety Bulb, Electron Microscopy Sciences
Safety Bulb
$56.90
Each
Natural rubber bulb allows safe, manual control of filling or discharging toxic liquids through a pipette by finger pressure, thus avoiding mouth contact.
  • Guaranteed to be consistently flexible
  • Désignation:
    Safety Bulb