Flat Embedding Molds, Electron Microscopy Sciences
Product Details & Documents
Single Tapered Ends have standard flat embedding mold, 21 numbered cavities, and measures L x W: 14 x 5 mm.
A clear silicone rubber with very high tear strength. It can be reused more times than the conventional silicone molds. It is resistant to most epoxy resins and its clear base allows positioning of the specimen lighted from beneath.
Single Tapered Ends have standard flat embedding mold, 21 numbered cavities, and measures L x W: 14 x 5 mm.
Small Flat Embedding Mold is made from clear silicone, the overall measurements are 1.65 x 3” with 12 cavities. Each cavity measures L x W x D: 10 x 6 x 4 mm (0.427 x 0.25 x 0.156”). This mold produces 12 embedding blocks with tapered ends.
Double Tapered Ends have a standard flat embedding mold where both ends are tapered. This enables you to embed specimens in both ends of each casting. Made from white silicone, 21 numbered cavities, each cavity measuring L x W x D: 14 x 4 x 4 mm.
Triple Tapered Ends have a new design with 10 triple numbered, tapered tip cavities to save you time and handling. Holds firmly in a chuck during trimming or sectioning. Made from white silicone, each cavity measures 8 mm from tip to center and 5 mm deep.
Small Flat Embedding Mold is made from clear silicone, the overall measurements are 1.65 x 3” with 12 cavities. Each cavity measures L x W x D: 10 x 6 x 4 mm (0.427 x 0.25 x 0.156”). This mold produces 12 embedding blocks with tapered ends.
Double Tapered Ends have a standard flat embedding mold where both ends are tapered. This enables you to embed specimens in both ends of each casting. Made from white silicone, 21 numbered cavities, each cavity measuring L x W x D: 14 x 4 x 4 mm.
Triple Tapered Ends have a new design with 10 triple numbered, tapered tip cavities to save you time and handling. Holds firmly in a chuck during trimming or sectioning. Made from white silicone, each cavity measures 8 mm from tip to center and 5 mm deep.
Spécifications
- Désignation:Flat Embed. Mold Triple Ends