Flat Embedding Molds, Electron Microscopy Sciences | Avantor
Flat Embedding Molds, Electron Microscopy Sciences
Flat Embed. Mold Triple Ends
Catalog # 100491-670
Fournisseur #:  70875
Flat Embedding Molds, Electron Microscopy Sciences
Catalog # 100491-670
Flat Embedding Molds, Electron Microscopy Sciences
Flat Embedding Molds, Electron Microscopy Sciences
Flat Embed. Mold Triple Ends
Price
$50.52
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Single Tapered Ends have standard flat embedding mold, 21 numbered cavities, and measures L x W: 14 x 5 mm.
A clear silicone rubber with very high tear strength. It can be reused more times than the conventional silicone molds. It is resistant to most epoxy resins and its clear base allows positioning of the specimen lighted from beneath.
Single Tapered Ends have standard flat embedding mold, 21 numbered cavities, and measures L x W: 14 x 5 mm.
Small Flat Embedding Mold is made from clear silicone, the overall measurements are 1.65 x 3” with 12 cavities. Each cavity measures L x W x D: 10 x 6 x 4 mm (0.427 x 0.25 x 0.156”). This mold produces 12 embedding blocks with tapered ends.
Double Tapered Ends have a standard flat embedding mold where both ends are tapered. This enables you to embed specimens in both ends of each casting. Made from white silicone, 21 numbered cavities, each cavity measuring L x W x D: 14 x 4 x 4 mm.
Triple Tapered Ends have a new design with 10 triple numbered, tapered tip cavities to save you time and handling. Holds firmly in a chuck during trimming or sectioning. Made from white silicone, each cavity measures 8 mm from tip to center and 5 mm deep.
Small Flat Embedding Mold is made from clear silicone, the overall measurements are 1.65 x 3” with 12 cavities. Each cavity measures L x W x D: 10 x 6 x 4 mm (0.427 x 0.25 x 0.156”). This mold produces 12 embedding blocks with tapered ends.
Double Tapered Ends have a standard flat embedding mold where both ends are tapered. This enables you to embed specimens in both ends of each casting. Made from white silicone, 21 numbered cavities, each cavity measuring L x W x D: 14 x 4 x 4 mm.
Triple Tapered Ends have a new design with 10 triple numbered, tapered tip cavities to save you time and handling. Holds firmly in a chuck during trimming or sectioning. Made from white silicone, each cavity measures 8 mm from tip to center and 5 mm deep.
Spécifications
  • Désignation:
    Flat Embed. Mold Triple Ends
Certains produits peuvent apparaître comme restreints
Pour garantir un passage en caisse fluide et rapide,veuillez-vous connecter à votre compte.Certains articles peuvent apparaître comme restreints simplement parce que vous n'êtes pas connecté.
Si vous n'avez pas de compte, vous pouvez vous inscrire en utilisant notre formulaire d'inscription en ligne (https://www.avantorsciences.com/ca/en/login/register)
Si des restrictions apparaissent toujours après votre connexion, certains produits — comme les produits chimiques ou les dispositifs médicaux — nécessitent des étapes supplémentaires de vérification de compte pour pouvoir passer une commande. Certains articles peuvent également nécessiter une licence spécifique ou une documentation supplémentaire ; la documentation requise sera demandée pour ces articles avant l’expédition.
Recommandations
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.