Colormark ™; Plus Glass Slide, Electron Microscopy Sciences | Avantor
Colormark ™; Plus Glass Slide, Electron Microscopy Sciences
Colormark slides, colored ends, Green Front & Back, 1 g
Catalog # 100491-378
Fournisseur #:  63400-30
Colormark ™; Plus Glass Slide, Electron Microscopy Sciences
Catalog # 100491-378
Supplier:  
Colormark ™; Plus Glass Slide, Electron Microscopy Sciences
Colormark ™; Plus Glass Slide, Electron Microscopy Sciences
Colormark slides, colored ends, Green Front & Back, 1 g
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Specimen slide identification is vital automated etched slide labeling instruments ensure specimen slide integrity but are difficult to read on ordinary slides.
To clearly read diamond scribed slide labels against any surface, you need the only slide designed to be used with your automated slide labeling instrument.
You need COLORMARK slides.These brightly printed slides greatly improve etched labeling visibility and offer all the advantages of Superfrost slides. A bright contrasting color has been printed on the back of the Superfrost end so that patient or specimen information etched through the painted end on the front is clearly visible. Colormark Plus slides are adhesive coated therefore ensuring the adhesion of the section to the slide during staining procedures.
Spécifications
  • Capacité:
    1 g
  • Color:
    Green Front & Back
  • Désignation:
    Colormark slides, colored ends