Emery Paper, Electron Microscopy Sciences | Avantor
Emery Paper, Electron Microscopy Sciences
Emery paper, 3 grit, 180 mesh
Catalog # 100490-140
Emery Paper, Electron Microscopy Sciences
Catalog # 100490-140
Supplier:  
Emery Paper, Electron Microscopy Sciences
Emery Paper, Electron Microscopy Sciences
Emery paper, 3 grit, 180 mesh
Discontinued
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This emery paper comes in sheets and in varying grit sizes.
Grit 4/0 is the finest and 3 is the coarsest.
Size: 22×34 cm (9×13¾")
Spécifications
  • Désignation:
    Emery paper, 3 grit, 180 mesh