Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Product Details & Documents
For collecting and exhibiting a wide variety of specimens in the lab or field. Caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suitable for hydroscopic materials.
Spécifications
- Capacité:32 ml
- Désignation:With Plastic Snap Cap
- Taille:32 ml