DrainRack™ Jr. Slide Rack, Electron Microscopy Sciences
Product Details & Documents
Made from high impact polystyrene. This model is similar to the standard DrainRack™ but can hold up to 100 microscope slides in 50 individual numbered slots.
The rack is autoclavable. Dimensions: 75 x 125 x 25 mm (3 x 5 x 1").
Spécifications
- Color:Green