Finger Cots, Latex, Electron Microscopy Sciences
Large
  100488-428
Finger Cots, Latex, Electron Microscopy Sciences
  100488-428
Finger Cots, Latex, Electron Microscopy Sciences
Finger Cots, Latex, Electron Microscopy Sciences
Large
$29.26
Each Pack of 144
Inexpensive, expandable finger cots.
  • Seamless
  • Protective dexterity without covering the entire hand
  • Powderless
  • Non-rolled
3 1/2 mil (0.0030"). Length 40 mm (2 3/4").
  • Taille:
    Large