COVAMAT® Lining, Electron Microscopy Sciences | Avantor
COVAMAT® Lining, Electron Microscopy Sciences
Covamat Lining, 50', 50 ft
Catalog # 100488-120
Fournisseur #:  72615-50
COVAMAT® Lining, Electron Microscopy Sciences
Catalog # 100488-120
COVAMAT® Lining, Electron Microscopy Sciences
COVAMAT® Lining, Electron Microscopy Sciences
Covamat Lining, 50', 50 ft
Price
$528.87
Log in pour la disponibilité
Each (50Feet) Roll of 50
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Versatile clear lining material: clear polyethylene ribbed matting can be used for lining laboratory bench top, shelves, trays, and more.
It is easily trimmed and also works well under drums or carboys. 0.25 mm (0.010") width 0.7 mm (0.030") thick ribs, 76 cm (30") wide, 15.2 m (50') and 4.6 m (15') roll. 1 roll/box.
Spécifications
  • Capacité:
    50 ft
  • Désignation:
    Covamat Lining, 50'