COVAMAT® Lining, Electron Microscopy Sciences
Product Details & Documents
Versatile clear lining material: clear polyethylene ribbed matting can be used for lining laboratory bench top, shelves, trays, and more.
It is easily trimmed and also works well under drums or carboys. 0.25 mm (0.010") width 0.7 mm (0.030") thick ribs, 76 cm (30") wide, 15.2 m (50') and 4.6 m (15') roll. 1 roll/box.
Spécifications
- Capacité:15 ft
- Désignation:Covamat Lining, 15'