Capsule Molds, Electron Microscopy Sciences
Product Details & Documents
Our molds are made from the strongest possible silicone rubber providing extremely high tear resistance. They are all reusable and produce embedding blocks of numerous shapes and sizes with a wide variety of tips. We also offer a mold which produces blocks to be used for re-embedding.
Spécifications
- Désignation:Capsule Mold Type A, 10 Cavities