Round Bottom Flasks
25 ml, 14/23, Round
Catalog # CA11009-144
Supplier: Heidolph NA, LLC
Supplier #: 015890750CA
Jump to:
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Spécifications
- ST Joint No.:14/23
- Capacité:25 ml
- Bottom Style:Round