Round Bottom Flasks
Round Bottom Flasks
25 ml, 14/23, Round
Catalog # CA11009-144
Supplier:   Heidolph NA, LLC
Supplier #:  015890750CA
Round Bottom Flasks
Catalog # CA11009-144
Supplier:  Heidolph NA, LLC
Round Bottom Flasks
25 ml, 14/23, Round
Price
$679.53
Log in pour la disponibilité
Pack of 2
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Spécifications
  • ST Joint No.:
    14/23
  • Capacité:
    25 ml
  • Bottom Style:
    Round