Round Bottom Flasks
5 ml, 14/23, Round
Catalog # CA11009-134
Supplier: Heidolph NA, LLC
Supplier #: 015890700CA
Jump to:
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Specifications
- ST Joint No.:14/23
- Capacité:5 ml
- Bottom Style:Round