Round Bottom Flasks
Round Bottom Flasks
5 ml, 14/23, Round
Catalog # CA11009-134
Supplier:   Heidolph NA, LLC
Supplier #:  015890700CA
Round Bottom Flasks
Catalog # CA11009-134
Supplier:  Heidolph NA, LLC
Round Bottom Flasks
5 ml, 14/23, Round
Price
$509.20
Log in for availability
Pack of 5
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Specifications
  • ST Joint No.:
    14/23
  • Capacité:
    5 ml
  • Bottom Style:
    Round