Round Bottom Flasks
10 ml, 14/23, Round
Catalog # CA11009-130
Supplier: Heidolph NA, LLC
: 015890680CA
Jump to:
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Spécifications
- ST Joint No.:14/23
- Capacité:10 ml
- Bottom Style:Round