Round Bottom Flasks
50 ml, 19/26, Round
  CA11009-124
 :  015890650CA
Round Bottom Flasks
  CA11009-124
Round Bottom Flasks
50 ml, 19/26, Round
$682.86
Pack of 5
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
  • ST Joint No.:
    19/26
  • Capacité:
    50 ml
  • Bottom Style:
    Round