Round Bottom Flasks
150 ml, 19/26, Round
Catalog # CA11009-116
Supplier:   Heidolph NA, LLC
 :  015890610CA
Round Bottom Flasks
Catalog # CA11009-116
Supplier:  Heidolph NA, LLC
Round Bottom Flasks
150 ml, 19/26, Round
Price
$818.22
Log in pour la disponibilité
Pack of 5
Product Details & Documents
These round bottom flasks are made up of glass, for use with StarFish™ multi-experiment heating and stirring work station.
Spécifications
  • ST Joint No.:
    19/26
  • Capacité:
    150 ml
  • Bottom Style:
    Round