VWR® CryoPro® Plastic Storage Boxes
2" High Mini Cryogenic Freezer Storage Box, 25, 7.6L×5.1H×7.6D cm (3×2×3")
Catalog # 89214-762
Supplier: VWR International
Jump to:
Product Details & Documents
Polycarbonate construction for use in mechanical and LN2 freezers.
- Grid format for lids
- Boxes feature drain slots for safe liquid nitrogen storage
Lids come printed with 25, 81, and 100 cell grids to assist with inventory control.
Specifications
- Nb de places:25
- Désignation:2" High Mini Cryogenic Freezer Storage Box
- Dimensions:7.6L×5.1H×7.6D cm (3×2×3")