VWR® CryoPro® Plastic Storage Boxes
VWR® CryoPro® Plastic Storage Boxes
2" High Mini Cryogenic Freezer Storage Box, 25, 7.6L×5.1H×7.6D cm (3×2×3")
Catalog # 89214-762
Supplier:   VWR International
VWR® CryoPro® Plastic Storage Boxes
Catalog # 89214-762
Supplier:  VWR International
VWR® CryoPro® Plastic Storage Boxes
VWR® CryoPro® Plastic Storage Boxes
2" High Mini Cryogenic Freezer Storage Box, 25, 7.6L×5.1H×7.6D cm (3×2×3")
Price
$25.54
New Customer Promotional Price
Log in for availability
Each Case of 50
Product Details & Documents
Polycarbonate construction for use in mechanical and LN2 freezers.
  • Grid format for lids
  • Boxes feature drain slots for safe liquid nitrogen storage
Lids come printed with 25, 81, and 100 cell grids to assist with inventory control.
Specifications
  • Nb de places:
    25
  • Désignation:
    2" High Mini Cryogenic Freezer Storage Box
  • Dimensions:
    7.6L×5.1H×7.6D cm (3×2×3")