VWR® CryoPro® Plastic Storage Boxes
2" High Large Cryogenic Freezer Storage Box, 81, 13.3L×5.1H×13.3D cm (51/4×2×51/4")
Catalog # 89214-746
Jump to:
Discontinued
Le produit n'est plus fabriqué. Cependant, nous avons trouvé des produits alternatifs similaires.
Alternatives
VWR BOX HIGH LG F/VIAL STORAGE 5X5X2 Catalog #89214-760
Product 89214-760 is similar to 89214-746 but manufacturer and specifications may vary.
BOX CRYO VWR 81 PLACE PCA BLUE 1-2ML PK5 Catalog #89236-046
Product 89236-046 is similar to 89214-746 but manufacturer and specifications may vary.
BOX CRYO VWR 81 PLC PCA MIX CR 1-2ML PK5 Catalog #89236-044
Product 89236-044 is similar to 89214-746 but manufacturer and specifications may vary.
Product Details & Documents
Polycarbonate construction for use in mechanical and LN2 freezers.
- Grid format for lids
- Boxes feature drain slots for safe liquid nitrogen storage
Lids come printed with 25, 81, and 100 cell grids to assist with inventory control.
Spécifications
- Nb de places:81
- Désignation:2" High Large Cryogenic Freezer Storage Box
- Dimensions:13.3L×5.1H×13.3D cm (51/4×2×51/4")



