SpectraMax® M5/M5e Multimode Plate Reader, Molecular Devices | Avantor
SpectraMax® M5/M5e Multimode Plate Reader, Molecular Devices
SpectraMax® M5e Microplate Reader
Catalog # 89212-400
Supplier:   Molecular Devices
Fournisseur #:  M5E
SpectraMax® M5/M5e Multimode Plate Reader, Molecular Devices
Catalog # 89212-400
Supplier:  Molecular Devices
SpectraMax® M5/M5e Multimode Plate Reader, Molecular Devices
SpectraMax® M5/M5e Multimode Plate Reader, Molecular Devices
SpectraMax® M5e Microplate Reader
Price
Renseignez-vous sur le prix
Log in pour la disponibilité
Each
Product Details & Documents
The SpectraMax® M5 Multi-Mode Microplate Reader delivers single mode reader performance and can be equipped to read volumes as low as 2uL in one multimode reader package.
  • Wide range of applications
  • Five preferred detection modes
  • Maximum signal with lowest background
The dual monochromator optics allow the widest range of applications to be utilized for bioresearch and drug discovery applications, all without the need to change filters
For fluorescence intensity, time resolved fluorescence, and fluorescence polarization assays, the SpectraMax M5 Microplate Reader optical design provides the highest level of flexibility. Users can select from top or bottom read modes for improved sensitivity for solution and cell-based assays. Assays can be better optimized by scanning across a range of wavelengths in increments as small as 1 nm. Up to 4 wavelength pairs can be read in one protocol for endpoint and kinetic measurements, allowing for fast setup of FRET and TR-FRET assays.
For luminescence, the SpectraMax® M5 Microplate Reader utilizes a dedicated luminescence photomultiplier tube (PMT), providing the user the maximum signal and lowest background possible for glow luminescence reporter gene assays.
The SpectraMax® M5 utilizes a single monochromator design for UV/VIS Absorbance, allowing the user to maximize sensitivity and tunability, and also uses patented PathCheck® Pathlength Measurement Technology: the only temperature independent pathlength correction for microplates. The M5 can read volumes down to 2uL with the uMax Low Volume Plate.
Ordering information: Includes current version of SoftMax Pro Software. Additional models are available. Additional accessories are available including the µMax Low Volume Plate, standard validation tools, and 21 CFR Part 11 Compliance tools. The SpectraMax® M5e includes htrf certification, and both the M5 and M5e are lanthascreen certified. The new Molecular Devices instrument comes with a standard 1-year depot warranty service. Purchase of Additional 1 Year Service Plan includes: MRC Essential Service Plan, including parts, labor, and shipping/handling costs for emergency repairs for one year at the Molecular Devices Repair Center. Priority repair scheduling along with unlimited technical support and loaner instrument (if available) are provided with this service option.
Spécifications
  • Désignation:
    SpectraMax® M5e Microplate Reader