Safety Containers
Waste disposal, 7.5 L (2 gal.), 2 gal.
  89212-348
 :   Justrite
 :  12165
Safety Containers
  89212-348
 :  Justrite
Safety Containers
Safety Containers
Waste disposal, 7.5 L (2 gal.), 2 gal.
$1,507.85
Each
Safely contain hazardous liquids and control vapors to reduce the risk of fire, protecting both personnel and facilities.
  • Safely contain hazardous liquids and control vapors to reduce the risk of fire
  • Helps comply with OSHA regulations and state/local fire codes
Cease-Fire® waste receptacles provide protection against trash fires.
Helps comply with OSHA regulations and state and local fire codes.
Improve efficiency with ease-of-use features which enhance pouring and filling operations.
  • Capacité:
    2 gal.
  • Désignation:
    Waste disposal, 7.5 L (2 gal.)
  • Certification:
    FM
Accessories
  •  
    Description
  • 56610-471
    Extinguishing Head
    Each - $168.17
  •  
    Description
  • 14222-224
    Tubing, 3/8", Stainless Steel, EDPM Seals
    Each - $1,444.62
  •  
    Description
  • 89087-386
    Disconnect Kit Replacement Stainless Steel
    Each - $1,375.83
  •  
    Description
  • 89087-388
    Disconnect Kit Replacement Polypropylene
    Each - $352.90
  •  
    Description
  • 89178-228
    Stainless Steel Disconnect Replacement For 12018
    Each - $1,538.56
  •  
    Description
  • 71990-050
    Funnelspout Safety Can
    Each - $38.42
  •  
    Description
  • 71990-054
    Hose Galvanized Safety Can
    Each - $186.40
  •  
    Description
  • 14222-220
    Tubing, 3/8", Stainless Steel, VITON Seals
    Each - $1,444.72
  •  
    Description
  • 14222-214
    Disconnect Assembly Stainless Steel, 1/4" Fitting
    Each - $1,016.71
  •  
    Description
  • 14222-216
    Tubing, 3/8", Stainless Steel, PTFE Seals
    Each - $1,258.01
  •  
    Description
  • 14222-222
    Tubing, 1/4", 3/8"
    Each - $1,444.62
  •  
    Description
  • 89212-342
    Spill Basin Hplc Can
    Each - $556.16