Safety Containers
Waste disposal, 7.5 L (2 gal.), 2 gal.
89212-348
: Justrite
: 12165
Safely contain hazardous liquids and control vapors to reduce the risk of fire, protecting both personnel and facilities.
- Safely contain hazardous liquids and control vapors to reduce the risk of fire
- Helps comply with OSHA regulations and state/local fire codes
Cease-Fire® waste receptacles provide protection against trash fires.
Helps comply with OSHA regulations and state and local fire codes.
Improve efficiency with ease-of-use features which enhance pouring and filling operations.
- Capacité:2 gal.
- Désignation:Waste disposal, 7.5 L (2 gal.)
- Certification:FM
Accessories
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description
- Description