Rectangular Low Profile Plastic Cup Sinks, Scientific Plastics
Rectangular Cup Sink Low Profile with Extended Tailpiece, White
89209-730
: F164L081-158
Rectangular Cup sink Ideal for retro-fit applications and ADA low countertop fumehoods.
- Beveled sides help reduce splash
- Tapered bottom eliminates water pooling
Regular Cup Skin for mounting at the rear of the fume hood and Extended tailpiece allows for mounting near the front of the fume hood.
- Color:White
- Désignation:Rectangular Cup Sink Low Profile with Extended Tailpiece