Rectangular Low Profile Plastic Cup Sinks, Scientific Plastics
Rectangular Cup Sink Low Profile, Grey
  89209-724
 :  W39081-154
Rectangular Low Profile Plastic Cup Sinks, Scientific Plastics
  89209-724
Rectangular Low Profile Plastic Cup Sinks, Scientific Plastics
Rectangular Low Profile Plastic Cup Sinks, Scientific Plastics
Rectangular Cup Sink Low Profile, Grey
$458.65
Each
Rectangular Cup sink Ideal for retro-fit applications and ADA low countertop fumehoods.
  • Beveled sides help reduce splash
  • Tapered bottom eliminates water pooling
Regular Cup Skin for mounting at the rear of the fume hood and Extended tailpiece allows for mounting near the front of the fume hood.
  • Color:
    Grey
  • Désignation:
    Rectangular Cup Sink Low Profile