Rectangular Low Profile Plastic Cup Sinks, Scientific Plastics
Rectangular Cup Sink Low Profile, Black
Catalog # 89209-720
Supplier: Scientific Plastics
Fournisseur #: W39081-150
Rectangular Low Profile Plastic Cup Sinks, Scientific Plastics
Catalog # 89209-720
Supplier: Scientific Plastics
Jump to:
Product Details & Documents
Rectangular Cup sink Ideal for retro-fit applications and ADA low countertop fumehoods.
- Beveled sides help reduce splash
- Tapered bottom eliminates water pooling
Regular Cup Skin for mounting at the rear of the fume hood and Extended tailpiece allows for mounting near the front of the fume hood.
Spécifications
- Color:Black
- Désignation:Rectangular Cup Sink Low Profile